Orders LinkContactUsLinkComponent
Burettes, removable stopcock
Burettes, removable stopcock
PTFE-stophane
Katalog # 612-0376
Leverandør:   SciLabware
Burettes, removable stopcock
Katalog # 612-0376
Leverandør:  SciLabware
Burettes, removable stopcock
Burettes, removable stopcock
PTFE-stophane
Price
3.010,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Soda lime glass, class B, removable PTFE stopcock.
  • Flared top for safer filling
  • Blue enamel graduations
BS 846
Certifications: BS 846
Specifikationer
  • Beskrivelse:
    PTFE-stophane
  • Seq:
    06