Torches
Low flow quartz torch with 2,4 mm ID injector tube
Katalog # VARI2010090400
Leverandør: VARIAN
Gå til:
Product Details & Documents
These torches light easily and stay lit, reducing downtime. With even and stable plasma to deliver reproducible results.
Specifikationer
- Beskrivelse:Low flow quartz torch with 2,4 mm ID injector tube
- Seq:04
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Spare quartz torch bonnet for radial ICP-OESSpare quartz torch bonnet for radial ICP-OESVARI2010070790Log ind for tilgængelighed1 * 1 Items - 1.690,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Torch alignment tool for radial ICP-OES
Produktinformation
- Katalog #TilgængelighedPrice
- GazFit fittings for one-piece and semi-demountable ICP-OES torchesGazFit fittings for one-piece and semi-demountable ICP-OES torchesVARI7910051290Log ind for tilgængelighed1 * 1 Items - 1.990,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Transfer tube FEP for axial ICP
Produktinformation
- Katalog #TilgængelighedPrice
- Transfer tube FEP Sturman-Masters spray chamber for radial ICPTransfer tube FEP Sturman-Masters spray chamber for radial ICPVARI7910051490Log ind for tilgængelighed1 * 1 Items - 1.990,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Kit Torch Fittings
Produktinformation
- Katalog #TilgængelighedPrice
- Pre-optics cone for Liberty Series axial ICP-OESPre-optics cone for Liberty Series axial ICP-OESVARI3610007300Log ind for tilgængelighed1 * 1 Items - 8.550,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Esteele cleaning powder, for cleaning interface cone on axially-viewed ICP-OESEsteele cleaning powder, for cleaning interface cone on axially-viewed ICP-OESVARI8110002800Log ind for tilgængelighed1 * 1 Items - 544,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Spare semi-demountable quartz torch body for radial ICP-OESSpare semi-demountable quartz torch body for radial ICP-OESVARI2010072900Log ind for tilgængelighed1 * 1 Items - 11.040,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Quartz injector tube, 0,8 mm
Produktinformation
- Katalog #TilgængelighedPrice
- Spare semi-demountable quartz torch body for axial ICP-OESSpare semi-demountable quartz torch body for axial ICP-OESVARI2010083400Log ind for tilgængelighed1 * 1 Items - 11.040,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Quartz injector tube, 1,4 mm
Produktinformation
- Katalog #TilgængelighedPrice
- Injector holder kit, includes O-ringInjector holder kit, includes O-ringVARI9910057300Log ind for tilgængelighed1 * 1 Items - 1.530,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- O-rings kit