Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
1,2-Dichlorpropan ≥98%

1,2-Dichlorpropan ≥98%

Glass bottle for solids, 25 g
Katalog # L03083.14
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
1,2-Dichlorpropan ≥98%
Katalog # L03083.14
CAS nummer:  78-87-5
1,2-Dichlorpropan ≥98%
1,2-Dichlorpropan ≥98%

Glass bottle for solids, 25 g

CAS nummer:  78-87-5
Price
320,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 78-87-5 Molecular Formula: C3H6Cl2 Formula Weight: 112.99 Flash Point: 4°(39°F) Refractive Index: 1.4380 Odour: Sweet Melting Point: -100° Boiling Point: 95-97° Beilstein Reference: 1718880 Merck Reference: 14,7854 UN No.: 1279 MDL No.: MFCD00000868

Synonymer:  Propylene dichloride
  • CAS nummer:78-87-5
  • Formel:ClH₂CCH₂CH₂Cl
  • MDL nummer:MFCD00000868
  • Molecular Weight:112.99 g/mol
  • Merck Index:14,7854
  • Kogende Pt:95…97 °C
  • Smeltende Pt:–100 °C
  • Tæthed:1,157
  • Flash Pt:4 °C (39 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-152-2
  • FN:1279
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.