Dipropylamin 99%
Glass bottle, 500 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 142-84-7 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: 7°(45°F) Refractive Index: 1.4050 Odour: Ammonia-like Melting Point: -63° Boiling Point: 109-110° Beilstein Reference: 505974 Merck Reference: 14,3343 UN No.: 2383 MDL No.: MFCD00009362
Synonymer: N-Propyl-1-propanamine
- CAS nummer:142-84-7
- Formel:C₆H₁₅N
- MDL nummer:MFCD00009362
- Molecular Weight:101.19 g/mol
- Merck Index:14,3343
- Kogende Pt:109…110 °C
- Smeltende Pt:–63 °C
- Tæthed:0,738
- Flash Pt:7 °C (45 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-565-9
- FN:2383
- ADR:3,II
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL



