Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Filter aid, Harborlite 800

500 g
Katalog # L13608.36
Irriterende
Leverandør:   Thermo Fisher Scientific
Filter aid, Harborlite 800
Katalog # L13608.36
CAS nummer:  
Filter aid, Harborlite 800
Filter aid, Harborlite 800

500 g

CAS nummer:  
Price
546,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

MDL No.: MFCD00197888

  • MDL nummer:MFCD00197888
  • Tæthed:2,3
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.