Methylethylketon ≥99%
Glass bottle, 500 mL
Leverandør: Thermo Fisher Scientific
Methylethylketon ≥99%
Katalog # L13185.AP
Leverandør: Thermo Fisher Scientific
CAS nummer: 78-93-3
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 78-93-3 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -3°(26°F) Refractive Index: 1.3785 Odour: Characteristic Melting Point: -87° Boiling Point: 80° Beilstein Reference: 741880 Merck Reference: 14,6072 UN No.: 1193 MDL No.: MFCD00011648
Synonymer: Ethylmethylketon, Butanon
- CAS nummer:78-93-3
- Formel:C₂H₅COCH₃
- MDL nummer:MFCD00011648
- Molecular Weight:72.11 g/mol
- Merck Index:14,6072
- Kogende Pt:80 °C
- Smeltende Pt:–87 °C
- Tæthed:0,805
- Flash Pt:–3 °C (26 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-159-0
- FN:1193
- ADR:3,II
- NÅ:01-2119457290-43
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
- Cat. No.:L13185.AP
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



