Natriumcarbonat ≥98%
Glass bottle for solids, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 98% CAS No.: 497-19-8 Molecular Formula: Na2CO3 Formula Weight: 105.99 Refractive Index: 1.535 Odour: Odourless pH: 11,5 Beilstein Reference: 4154566 Merck Reference: 14,8596 MDL No.: MFCD00003494
Synonymer: Calcined soda, Carbonic acid disodium salt
- CAS nummer:497-19-8
- Formel:Na₂CO₃
- MDL nummer:MFCD00003494
- Molecular Weight:105.99 g/mol
- Merck Index:14,8596
- Kogende Pt:1600 °C (1013 hPa)
- Smeltende Pt:854 °C
- Tæthed:2,53
- Opbevaringstemperatur:Stue temperatur
- EINECS:207-838-8
- NÅ:01-2119485498-19
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g
- Cat. No.:L13098.36
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



