Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Hexadecan ≥95%

n-Hexadecan ≥95%

1 gal US
Katalog # 43283.LV
Sundhed
Irriterende
Leverandør:   Thermo Fisher Scientific
n-Hexadecan ≥95%
Katalog # 43283.LV
CAS nummer:  544-76-3
n-Hexadecan ≥95%
n-Hexadecan ≥95%

1 gal US

CAS nummer:  544-76-3
Price
2.820,00 kr
Log ind for tilgængelighed
1 * 1 gal us
Product Details & Documents

Purity: 95% CAS No.: 544-76-3 Molecular Formula: CH3(CH2)14CH3 Formula Weight: 226.45 Flash Point: 135°(275°F) Physical Form: Liquid Density: 0.773 Refractive Index: 1.4345 Appearance: Colourless Melting Point: 18° Boiling Point: 287° Solubility: Soluble in ethanol Beilstein Reference: 1736592 MDL No.: MFCD00008998

Standard for rating diesel-fuel ignition qualities

Synonymer:  Hexadecane, Cetane
  • CAS nummer:544-76-3
  • Formel:H₃C(CH₂)₁₄CH₃
  • MDL nummer:MFCD00008998
  • Molecular Weight:226.45 g/mol
  • Kogende Pt:287 °C
  • Smeltende Pt:18 °C
  • Tæthed:0,773
  • Flash Pt:135 °C (275 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-878-9
Specifikationer

Specifikationer

  • Pakn:
    1 gal US

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.