Orders LinkContactUsLinkComponent
Ninhydrin ACS
Ninhydrin ACS
Glass bottle for solids, 500 g
Katalog # 43846.36
Irriterende
Leverandør:   Thermo Fisher Scientific
Ninhydrin ACS
Katalog # 43846.36
CAS nummer:  485-47-2
Ninhydrin ACS
Ninhydrin ACS
Glass bottle for solids, 500 g
CAS nummer:  485-47-2
Price
9.590,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
CAS No.: 485-47-2 Molecular Formula: C9H6O4 Formula Weight: 178.14 Physical Form: Crystalline Sensitivity: Light Sensitive Appearance: Pale yellow to pale brown Melting Point: 251° dec. Solubility: Soluble in water and alcohol Beilstein Reference: 1910963 Merck Reference: 14,6554 MDL No.: MFCD00003791
Reagent for determination of amino acids and amine containing compounds, as an intermediate.
Synonymer:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS nummer:485-47-2
  • Formel:C₉H₆O₄
  • MDL nummer:MFCD00003791
  • Molecular Weight:178.14 g/mol
  • Merck Index:14,6554
  • Kogende Pt:351 °C (1013 hPa)
  • Smeltende Pt:251 °C (dec.)
  • Tæthed:0,862 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:207-618-1
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g
Specifikationer