Triethylcitrat ≥99%
Glass bottle for solids, 250 g
Purity: 99% CAS No.: 77-93-0 Molecular Formula: C12H20O7 Formula Weight: 276.29 Flash Point: 176°(348°F) Refractive Index: 1.4420 Melting Point: -55° Boiling Point: 126-127°/1mm Beilstein Reference: 1801199 Merck Reference: 14,2326 MDL No.: MFCD00009201
: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester
- :77-93-0
- :C₁₂H₂₀O₇
- :MFCD00009201
- :276.29 g/mol
- :14,2326
- :126…127 °C (1 mmHg)
- :–55 °C
- :1,137
- :176 °C (348 °F)
- :Stue temperatur
- :201-070-7
- Pakning:Glass bottle for solids
- Pakn:250 g