Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
6-Methoxyindol ≥98%

6-Methoxyindol ≥98%

Glass bottle for solids, 1 g
Katalog # L19074.03
Irriterende
Leverandør:   Thermo Fisher Scientific
6-Methoxyindol ≥98%
Katalog # L19074.03
CAS nummer:  3189-13-7
6-Methoxyindol ≥98%
6-Methoxyindol ≥98%

Glass bottle for solids, 1 g

CAS nummer:  3189-13-7
Price
1.620,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Purity: 98+% CAS No.: 3189-13-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 90-94° Boiling Point: 105°/0.2mm MDL No.: MFCD00022780

Synonymer:  6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether
  • CAS nummer:3189-13-7
  • Formel:C₉H₉NO
  • MDL nummer:MFCD00022780
  • Molecular Weight:147.18 g/mol
  • Kogende Pt:105 °C (0,2 mmHg)
  • Smeltende Pt:90…94 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:221-689-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer