Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pentylacetat ≥99%

Pentylacetat ≥99%

Glass bottle for solids, 100 g
Katalog # L10852.22
Brandfarlig
Leverandør:   Thermo Fisher Scientific
Pentylacetat ≥99%
Katalog # L10852.22
CAS nummer:  628-63-7
Pentylacetat ≥99%
Pentylacetat ≥99%

Glass bottle for solids, 100 g

CAS nummer:  628-63-7
Price
332,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 628-63-7 Molecular Formula: CH3CO2(CH2)4CH3 Formula Weight: 130.19 Flash Point: 24°(75°F) Refractive Index: 1.4030 Odour: Fruit-like Melting Point: -100° Boiling Point: 142° Beilstein Reference: 1744753 UN No.: 1104 MDL No.: MFCD00009500

Synonymer:  n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate
  • CAS nummer:628-63-7
  • Formel:C₇H₁₄O₂
  • MDL nummer:MFCD00009500
  • Molecular Weight:130.19 g/mol
  • Kogende Pt:142 °C
  • Smeltende Pt:–100 °C
  • Tæthed:0,876
  • Flash Pt:24 °C (75 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:211-047-3
  • FN:1104
  • ADR:3,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.