Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Ethylbenzen ≥99%

Glass bottle, 500 mL
Katalog # L05908.AP
Brandfarlig
Sundhed
Irriterende
Leverandør:   Thermo Fisher Scientific
Ethylbenzen ≥99%
Katalog # L05908.AP
CAS nummer:  100-41-4
Ethylbenzen ≥99%
Ethylbenzen ≥99%

Glass bottle, 500 mL

CAS nummer:  100-41-4
Price
371,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Purity: 99% CAS No.: 100-41-4 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 15°(59°F) Refractive Index: 1.4950 Odour: Petrol-like Melting Point: -95° Boiling Point: 135-136° Beilstein Reference: 1901871 Merck Reference: 14,3765 UN No.: 1175 MDL No.: MFCD00011647

Synonymer:  Phenylethane
  • CAS nummer:100-41-4
  • Formel:C₈H₁₀
  • MDL nummer:MFCD00011647
  • Molecular Weight:106.17 g/mol
  • Merck Index:14,3765
  • Kogende Pt:135…136 °C
  • Smeltende Pt:–95 °C
  • Tæthed:0,867
  • Flash Pt:15 °C (59 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-849-4
  • FN:1175
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer