Orders LinkContactUsLinkComponent
Ethylbenzen ≥99%
Glass bottle, 100 mL
  L05908.AE
Ethylbenzen ≥99%
  L05908.AE
 :  100-41-4
Ethylbenzen ≥99%
Ethylbenzen ≥99%
Glass bottle, 100 mL
 :  100-41-4
328,00 kr
1 * 100 mL
Purity: 99% CAS No.: 100-41-4 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 15°(59°F) Refractive Index: 1.4950 Odour: Petrol-like Melting Point: -95° Boiling Point: 135-136° Beilstein Reference: 1901871 Merck Reference: 14,3765 UN No.: 1175 MDL No.: MFCD00011647
 :  Phenylethane
  •  :100-41-4
  •  :C₈H₁₀
  •  :MFCD00011647
  •  :106.17 g/mol
  •  :14,3765
  •  :135…136 °C
  •  :–95 °C
  •  :0,867
  •  :15 °C (59 °F)
  •  :Stue temperatur
  •  :202-849-4
  •  :1175
  •  :3,II
  • Pakning:
    Glass bottle
  • Pakn:
    100 mL