Ethylbenzen ≥99%
Glass bottle, 100 mL
Purity: 99% CAS No.: 100-41-4 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 15°(59°F) Refractive Index: 1.4950 Odour: Petrol-like Melting Point: -95° Boiling Point: 135-136° Beilstein Reference: 1901871 Merck Reference: 14,3765 UN No.: 1175 MDL No.: MFCD00011647
: Phenylethane
- :100-41-4
- :C₈H₁₀
- :MFCD00011647
- :106.17 g/mol
- :14,3765
- :135…136 °C
- :–95 °C
- :0,867
- :15 °C (59 °F)
- :Stue temperatur
- :202-849-4
- :1175
- :3,II
- Pakning:Glass bottle
- Pakn:100 mL