Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Nonadecan ≥99%

n-Nonadecan ≥99%

Glass bottle for solids, 25 g
Katalog # L05701.14
Leverandør:   Thermo Fisher Scientific
n-Nonadecan ≥99%
Katalog # L05701.14
CAS nummer:  629-92-5
n-Nonadecan ≥99%
n-Nonadecan ≥99%

Glass bottle for solids, 25 g

CAS nummer:  629-92-5
Price
479,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 629-92-5 Molecular Formula: CH3(CH2)17CH3 Formula Weight: 268.53 Flash Point: 168°(334°F) Refractive Index: 1.4410 Melting Point: 32-34° Boiling Point: 330-331° Beilstein Reference: 1742892 MDL No.: MFCD00009012

Synonymer:  N-nonadecane
  • CAS nummer:629-92-5
  • Formel:H₃C(CH₂)₁₇CH₃
  • MDL nummer:MFCD00009012
  • Molecular Weight:268.53 g/mol
  • Kogende Pt:330…331 °C
  • Smeltende Pt:32…34 °C
  • Tæthed:0,786
  • Flash Pt:168 °C (334 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:211-116-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.