Filter Holder Accessories
Swinnex 25 mm with back-pressure screen, polypropylene housing, female Luer-Lok® inlet connection male Luer outlet slip
Katalog # MILFXX6504707
Leverandør: Merck
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Swinnex 25 mm with back-pressure screen, polypropylene housing, female Luer-Lok® inlet connection male Luer outlet slip
- Seq:08