Erucic acid tech. 90%, Technical Grade
Glass bottle for solids, 250 g
Purity: 90% CAS No.: 112-86-7 Molecular Formula: C22H42O2 Formula Weight: 338.58 Flash Point: 110°(230°F) Melting Point: 29-33° Boiling Point: 265°/15mm Beilstein Reference: 1728049 Merck Reference: 14,3674 MDL No.: MFCD00063188
: (Z)-Docos-13-enoic acid
- :112-86-7
- :C₂₂H₄₂O₂
- :MFCD00063188
- :338.58 g/mol
- :14,3674
- :265 °C (15 mmHg)
- :29…33 °C
- :>110 °C (230 °F)
- :Stue temperatur
- :204-011-3
- Pakning:Glass bottle for solids
- Pakn:250 g