Orders LinkContactUsLinkComponent
Erucic acid tech. 90%, Technical Grade
Glass bottle for solids, 250 g
  L08685.30
Erucic acid tech. 90%, Technical Grade
  L08685.30
 :  112-86-7
Erucic acid tech. 90%, Technical Grade
Erucic acid tech. 90%, Technical Grade
Glass bottle for solids, 250 g
 :  112-86-7
1.250,00 kr
1 * 250 g
Purity: 90% CAS No.: 112-86-7 Molecular Formula: C22H42O2 Formula Weight: 338.58 Flash Point: 110°(230°F) Melting Point: 29-33° Boiling Point: 265°/15mm Beilstein Reference: 1728049 Merck Reference: 14,3674 MDL No.: MFCD00063188
 :  (Z)-Docos-13-enoic acid
  •  :112-86-7
  •  :C₂₂H₄₂O₂
  •  :MFCD00063188
  •  :338.58 g/mol
  •  :14,3674
  •  :265 °C (15 mmHg)
  •  :29…33 °C
  •  :>110 °C (230 °F)
  •  :Stue temperatur
  •  :204-011-3
  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g