Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Erucic acid tech. 90%, Technical Grade

Erucic acid tech. 90%, Technical Grade

Glass bottle for solids, 50 g
Katalog # L08685.18
Leverandør:   Thermo Fisher Scientific
Erucic acid tech. 90%, Technical Grade
Katalog # L08685.18
CAS nummer:  112-86-7
Erucic acid tech. 90%, Technical Grade
Erucic acid tech. 90%, Technical Grade

Glass bottle for solids, 50 g

CAS nummer:  112-86-7
Price
385,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

Purity: 90% CAS No.: 112-86-7 Molecular Formula: C22H42O2 Formula Weight: 338.58 Flash Point: 110°(230°F) Melting Point: 29-33° Boiling Point: 265°/15mm Beilstein Reference: 1728049 Merck Reference: 14,3674 MDL No.: MFCD00063188

Synonymer:  (Z)-Docos-13-enoic acid
  • CAS nummer:112-86-7
  • Formel:C₂₂H₄₂O₂
  • MDL nummer:MFCD00063188
  • Molecular Weight:338.58 g/mol
  • Merck Index:14,3674
  • Kogende Pt:265 °C (15 mmHg)
  • Smeltende Pt:29…33 °C
  • Flash Pt:>110 °C (230 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-011-3
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.