Orders LinkContactUsLinkComponent
Linolsyre 95%
Linolsyre 95%
Glass bottle for solids, 5 g
Katalog # L07949.06
Leverandør:   Thermo Fisher Scientific
Linolsyre 95%
Katalog # L07949.06
CAS nummer:  60-33-3
Linolsyre 95%
Linolsyre 95%
Glass bottle for solids, 5 g
CAS nummer:  60-33-3
Price
746,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Purity: 95% CAS No.: 60-33-3 Molecular Formula: C18H32O2 Formula Weight: 280.45 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4699 Odour: Weak, characteristic Melting Point: -12° Boiling Point: 230°/16mm Beilstein Reference: 1727101 Merck Reference: 14,5505 MDL No.: MFCD00064241
Synonymer:  (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
  • CAS nummer:60-33-3
  • Formel:C₁₈H₃₂O₂
  • MDL nummer:MFCD00064241
  • Molecular Weight:280.45 g/mol
  • Merck Index:14,5505
  • Kogende Pt:230 °C (16 mmHg)
  • Smeltende Pt:–12 °C
  • Tæthed:0,902
  • Flash Pt:>110 °C (230 °F)
  • Opbevaringstemperatur:Køleskab
  • EINECS:200-470-9
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g