Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Test kits, chlorine, Comparator 2000+

Test kits, chlorine, Comparator 2000+

Special chlorine test kit, -, AF 112A/E/F, 1 KIT
Katalog # LOVI411128
Leverandør:   Lovibond
Test kits, chlorine, Comparator 2000+
Katalog # LOVI411128
Leverandør:  Lovibond
CAS nummer:  
Test kits, chlorine, Comparator 2000+
Test kits, chlorine, Comparator 2000+

Special chlorine test kit, -, AF 112A/E/F, 1 KIT

CAS nummer:  
Price
7.000,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Comparator 2000+ is a versatile modular system for testing water. Applicable in pool-water, research centres, universities, special applications, laboratory and field testing.

  • Compact and portable
  • Provide accurate and reproducible results
  • Colour-stable, fade-free glass standards
  • More than 400 different test discs are available
  • Compensation for coloured and turbid samples
  • Guaranteed constancy of the coloured glass standards

The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.

Certifications: In accordance with ISO 7393/2 'Determination of free chlorine and total chlorine'.

Leveringsinformation: Kit is supplied in a robust carrying case containing comparator 2000+, chlorine comparator disc, 4 x comparator vials, 10ml, 13mm path length, tablet reagents, crushing/stirring rod, vial cleaning brush and instruction sheet.

Specifikationer

Specifikationer

  • Pakn:
    1 KIT
  • Beskrivelse:
    Special chlorine test kit
  • Model:
    AF 112A/E/F
  • Område:
    -

Specifikationer