Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Test kits, chlorine, Comparator 2000+

Test kits, chlorine, Comparator 2000+

Chlorine (free, combined, total) test kit, 0,02 - 0,3 mg/l;
0,2 - 0,8 mg/l (Cl2), AF 112E/F, 1 KIT
Catalog # LOVI411126
Supplier:   Lovibond
Test kits, chlorine, Comparator 2000+
Catalog # LOVI411126
Supplier:  Lovibond
CAS Number:  
Test kits, chlorine, Comparator 2000+
Test kits, chlorine, Comparator 2000+

Chlorine (free, combined, total) test kit, 0,02 - 0,3 mg/l;
0,2 - 0,8 mg/l (Cl2), AF 112E/F, 1 KIT

CAS Number:  
Price
11.520,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Comparator 2000+ is a versatile modular system for testing water. Applicable in pool-water, research centres, universities, special applications, laboratory and field testing.

  • Compact and portable
  • Provide accurate and reproducible results
  • Colour-stable, fade-free glass standards
  • More than 400 different test discs are available
  • Compensation for coloured and turbid samples
  • Guaranteed constancy of the coloured glass standards

The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.

Certifications: In accordance with ISO 7393/2 'Determination of free chlorine and total chlorine'.

Delivery information: Kit is supplied in a robust carrying case containing comparator 2000+, chlorine comparator disc, 4 x comparator vials, 10ml, 13mm path length, tablet reagents, crushing/stirring rod, vial cleaning brush and instruction sheet.

Specifications

Specifications

  • Pakn:
    1 KIT
  • Beskrivelse:
    Chlorine (free, combined, total) test kit
  • Model:
    AF 112E/F
  • Område:
    0,02 - 0,3 mg/l;<br />0,2 - 0,8 mg/l (Cl<sub>2</sub>)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.