Accessories for comparators
Chlorine, 3/2APH, 2,0-10,0 mg/l
Katalog # LOVI232060
Leverandør: Lovibond
Gå til:
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Specifikationer
- Område:2,0-10,0 mg/l
- Seq:18
- Kode:3/2APH
- Testmetode:Chlorine