Orders LinkContactUsLinkComponent
Accessories for comparators
Accessories for comparators
Iron, 3/144, 0,02-0,3 mg/l
Katalog # LOVI230380
Leverandør:   Lovibond
Accessories for comparators
Katalog # LOVI230380
Leverandør:  Lovibond
Accessories for comparators
Accessories for comparators
Iron, 3/144, 0,02-0,3 mg/l
Price
2.960,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Specifikationer
  • Område:
    0,02-0,3 mg/l
  • Seq:
    30
  • Kode:
    3/144
  • Testmetode:
    Iron