Orders LinkContactUsLinkComponent
Stoppers, with drip tip, MBL®
Stoppers, with drip tip, MBL®
14/23, With pointed bottom
Katalog # 217-0746
Leverandør:   SciLabware
Stoppers, with drip tip, MBL®
Katalog # 217-0746
Leverandør:  SciLabware
Stoppers, with drip tip, MBL®
Stoppers, with drip tip, MBL®
14/23, With pointed bottom
Price
542,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Ideally suited for use with volumetric flasks etc. Lower closed end of stopper features a 'drip-tip' to ensure adhered liquid is returned to the flask. Flat head allows stopper to be stood on end, thus preventing contamination of the ground surface.
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Hollow-blown to reduce weight
Specifikationer
  • NS:
    14/23
  • Model:
    With pointed bottom
  • Seq:
    04