Pouring rings
Pouring ring, GL 32, klar
Katalog # BIBB4516/32D
Leverandør: SciLabware
Gå til:
Product Details & Documents
Pouring rings, made of Polypropylene, for laboratory/media bottles.
- Ensures drip-free pouring
- Very good chemical resistance
- Autoclavable up to 140 °C
Specifikationer
- Gevind:GL 32
- Farve:klar
- Beskrivelse:Pouring ring
- Seq:01