Screw cap for reagent bottles
42 mm
Katalog # BIBB4513/42M
Leverandør: SciLabware
Gå til:
Product Details & Documents
Phenolic cap with PTFE faced sealing disc
- Fits PYREX® reagent bottles
- Excellent chemical resistance
- Autoclavable up to 121 °C
Specifikationer
- Gevind:42 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



