Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Gas dispersion tubes

1, 300 mm, 8 mm, 15 mm
  BIBB3830/02M
 :   SciLabware
Gas dispersion tubes
  BIBB3830/02M
Gas dispersion tubes
Gas dispersion tubes

1, 300 mm, 8 mm, 15 mm

 
1.100,00 kr
   
1 * 2 Items
 

These Pyrex® borosilicate glass tubes for the dispersion of gas in liquids with sintered thimble allow for the choice of porosities to achieve optimum gas bubble size.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion with relatively high resistance to temperature changes
 
  • Ø hoved:
    15 mm
  • Porøsitet:
    1
  • Rør udv. Ø:
    8 mm
  • Længde:
    300 mm