Orders LinkContactUsLinkComponent
Spare lids for dessicators
Spare lids for dessicators
PYREX® borosilicate glass lid, With socket size 24/29, for desiccators vacuum type, 200 DN
Katalog # BIBB1594/04D
Leverandør:   SciLabware
Spare lids for dessicators
Katalog # BIBB1594/04D
Leverandør:  SciLabware
Spare lids for dessicators
Spare lids for dessicators
PYREX® borosilicate glass lid, With socket size 24/29, for desiccators vacuum type, 200 DN
Price
2.900,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Lids are made of PYREX® borosilicate glass.
Specifikationer
  • Nominel størrelse:
    200 DN
  • Model:
    With socket size 24/29, for desiccators vacuum type
  • Beskrivelse:
    PYREX® borosilicate glass lid