Spare lids for dessicators
PYREX® borosilicate glass lid, With socket size 24/29, for desiccators vacuum type, 150 DN
Katalog # BIBB1593/04D
Leverandør: SciLabware
Gå til:
Product Details & Documents
Lids are made of PYREX® borosilicate glass.
Specifikationer
- Nominel størrelse:150 DN
- Model:With socket size 24/29, for desiccators vacuum type
- Beskrivelse:PYREX® borosilicate glass lid