Spherical ground joints, Rotulex®
joint size 41
Katalog # BIBBRF41
Leverandør: SciLabware
Gå til:
Product Details & Documents
Made of borosilicate glass.
- All glass and PTFE pathway is highly resistant to chemical attack
- Can be used at temperatures up to 200 °C
- System comprises male ball joint with PTFE covered 'O' ring which fits into a female cup joint
- Joint can be retained together with spring loaded metal clip
Rotulex® offers a grease free spherical joint.
: Component parts should be ordered separately .
Specifikationer
- Model:joint size 41
- Seq:09