Orders LinkContactUsLinkComponent
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
500 ml, 19/26 NS, 24/29 NS
Katalog # 532-0086
Leverandør:   SciLabware
Separating funnels, pear shaped, Quickfit®
Katalog # 532-0086
Leverandør:  SciLabware
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
500 ml, 19/26 NS, 24/29 NS
Price
1.410,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Excellent chemical resistance
  • Excellent temperature resistance
Funnel stem with cone to allow easy connection to Quickfit® flask with interchangeable PTFE key stopcock.
Certifications: Complies with ISO 4800 type 2.
Specifikationer
  • Konusstr.:
    19/26 NS
  • Kappestr.:
    24/29 NS
  • Kapacitet:
    500 ml
  • Seq:
    08