Orders LinkContactUsLinkComponent
Separating funnels, pear shaped, Quickfit®
100 ml, 19/26 NS, 19/26 NS
Katalog # 532-0083
Leverandør:   SciLabware
Separating funnels, pear shaped, Quickfit®
Katalog # 532-0083
Leverandør:  SciLabware
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
100 ml, 19/26 NS, 19/26 NS
Price
1.080,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Excellent chemical resistance
  • Excellent temperature resistance
Funnel stem with cone to allow easy connection to Quickfit® flask with interchangeable PTFE key stopcock.
 : Complies with ISO 4800 type 2.
Specifikationer
  • Konusstr.:
    19/26 NS
  • Kappestr.:
    19/26 NS
  • Kapacitet:
    100 ml
  • Seq:
    05