Orders LinkContactUsLinkComponent
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
25 ml, 14/23 NS, 14/23 NS
Katalog # 532-0079
Leverandør:   SciLabware
Separating funnels, pear shaped, Quickfit®
Katalog # 532-0079
Leverandør:  SciLabware
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
25 ml, 14/23 NS, 14/23 NS
Price
1.050,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Excellent chemical resistance
  • Excellent temperature resistance
Funnel stem with cone to allow easy connection to Quickfit® flask with interchangeable PTFE key stopcock.
Certifications: Complies with ISO 4800 type 2.
Specifikationer
  • Konusstr.:
    14/23 NS
  • Kappestr.:
    14/23 NS
  • Kapacitet:
    25 ml
  • Seq:
    01