Orders LinkContactUsLinkComponent
Dropping funnels, cylindrical, Quickfit®
Dropping funnels, cylindrical, Quickfit®
100 ml, 14/23 NS, 14/23 NS
Katalog # 532-0075
Leverandør:   SciLabware
Dropping funnels, cylindrical, Quickfit®
Katalog # 532-0075
Leverandør:  SciLabware
Dropping funnels, cylindrical, Quickfit®
Dropping funnels, cylindrical, Quickfit®
100 ml, 14/23 NS, 14/23 NS
Price
721,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Funnel stem with ground-glass cone for flasks and assemblies. Stopcock has interchangeable glass key.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Certifications: Complies with ISO 4800
Specifikationer
  • Konusstr.:
    14/23 NS
  • Kappestr.:
    14/23 NS
  • Kapacitet:
    100 ml
  • Seq:
    01