Powder funnels, Quickfit®
29/32 NS, 105 mm
Catalog # 532-0072
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, stem with cone.
- Meget god kemisk resistens
- Høj temperaturresistens
Designed for transfer of powders into flasks and vessels.
Specifications
- Konusstr.:29/32 NS
- Tragt udv. Ø:105 mm
- Seq:04