Orders LinkContactUsLinkComponent
Powder funnels, Quickfit®
Powder funnels, Quickfit®
29/32 NS, 105 mm
Catalog # 532-0072
Supplier:   SciLabware
Powder funnels, Quickfit®
Catalog # 532-0072
Supplier:  SciLabware
Powder funnels, Quickfit®
Powder funnels, Quickfit®
29/32 NS, 105 mm
Price
457,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, stem with cone.
  • Meget god kemisk resistens
  • Høj temperaturresistens
Designed for transfer of powders into flasks and vessels.
Specifications
  • Konusstr.:
    29/32 NS
  • Tragt udv. Ø:
    105 mm
  • Seq:
    04