Non return valves, Azlon®
6-8 mm ID
Katalog # AZLONWH902P
Leverandør: SciLabware
Gå til:
Product Details & Documents
Designed for use in vacuum lines and fluid handling systems.
- Useful for preventing back pressure in filter flasks
- Printed arrow indicates the direction of flow
- Rigid with stepped tapered arms to give firm grip on tubing
Specifikationer
- Slangestørrelse:6-8 mm ID