Swabs tubes
Almindelig podepind, bulk-pakning, Wood/cotton, +
Katalog # GREISWA5_1000
Leverandør: Greiner Bio-One
Gå til:
Product Details & Documents
Swab in tubes suitable for the collection and transport of non-human bacteriological, serological, and cytological samples.
- Sterilised by irradiation
- Swab tubes made of polystyrene
Useful for hygienic controls in veterinary research as well as in environmental sampling.
Packaging: Individually wrapped.
Caution: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifikationer
- Steril:+
- Beskrivelse:Almindelig podepind, bulk-pakning
- Materiale:Wood/cotton