Counting chambers, Bürker
Tællekamre, mørk kant, med clips, Bürker-Türk, 0,10 mm
631-0922
Glass counting chambers equipped with two sets of grids, engraved into two ground, and polished surfaces on the central ridge. This central ridge is located between two elevated ridges, that are also ground and polished.
Designed for investigations of cell suspensions.
Conform to DIN 12847 and the German Calibration Ordinance
: DIN 12847 and the German Calibration Ordinance.
: Supplied in a transparent box.
- Kammerdybde:0,10 mm
- Beskrivelse:Tællekamre, mørk kant, med clips, Bürker-Türk
- Seq:09