Counting chambers, Bürker
Cell counting chamber, dark lined, Bürker-Türk, 0,10 mm
Katalog # 630-1543
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Glass counting chambers equipped with two sets of grids, engraved into two ground, and polished surfaces on the central ridge. This central ridge is located between two elevated ridges, that are also ground and polished.
Designed for investigations of cell suspensions.
Conform to DIN 12847 and the German Calibration Ordinance
Certifications: DIN 12847 and the German Calibration Ordinance.
Leveringsinformation: Supplied in a transparent box.
Documents
Specifikationer
- Kammerdybde:0,10 mm
- Beskrivelse:Cell counting chamber, dark lined, Bürker-Türk
- Seq:07