Celletællere, Neubauer
Counting chamber, dark-lined, Neubauer, 0,10 mm
Katalog # MARI0640110
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
These counting chambers are designed for counting leucocytes and erythrocytes.
- Double net ruling
- Two counting grids with one division
Certifications: IVD 98/79/EC
Leveringsinformation: Supplied with two cover glasses.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Kammerdybde:0,10 mm
- Beskrivelse:Counting chamber, dark-lined, Neubauer
- Seq:03