Celletæller med tre gitre, McMaster
Tællekammer, 1,5 mm, 127×26 mm
Katalog # 631-0695
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
This cell counter is suitable for liquid media.
- Three counting grids and spare cover glass
- Pair of clamps to fix the cover glass
- Ground edges and rounded corners
Specifikationer
- Kammerdybde:1,5 mm
- B×D:127×26 mm
- Beskrivelse:Tællekammer