Counting chambers, Malassez
Cell counting chamber with dark lines, Malassez, 0,2 mm
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Counting chambers specially designed for in vitro diagnostic.
- Double net ruling
- Two cover glasses
- Dark lined without clamps
Documents
Specifikationer
- Kammerdybde:0,2 mm
- Model:Malassez
- Beskrivelse:Cell counting chamber with dark lines
- Seq:001