Sample vials, trident vials, squat or tall
Tall, 7,00 ml, 17×58 mm, Yes
Katalog # 212-7073
Leverandør: MURRAY S
Gå til:
Product Details & Documents
Vials made from neutral glass with PE plug stoppers. Suitable for a wide variety of uses in the laboratory, production, more.
- Robust and economical, will withstand moderate temperature change
- Wide opening for easy filling and emptying
- Plug stopper for secure closure
Packaging: For production related reasons, the number of vials per box may vary.
Specifikationer
- Kasket inkluderet:Yes
- Diameter×H:17×58 mm
- Kapacitet:7,00 ml
- Beskrivelse:Tall



