Orders LinkContactUsLinkComponent
Refraktometrer, Håndholdt, Eclipse
Refraktometrer, Håndholdt, Eclipse
Sukker, 45 - 80 °Bx, 0,2 °Bx
Catalog # 635-0293
Supplier:   BELLINGHAM STANLEY
Refraktometrer, Håndholdt, Eclipse
Catalog # 635-0293
Supplier:  BELLINGHAM STANLEY
Refraktometrer, Håndholdt, Eclipse
Refraktometrer, Håndholdt, Eclipse
Sukker, 45 - 80 °Bx, 0,2 °Bx
Price
3.670,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Handheld refractometers use high precision clear scale, and simple zero adjustment with lock.
  • Shaded field of view for distinction
  • Anti-roll support and ergonomic design
  • Comfortable eyepiece with smooth adjustment
  • Easy to clean prism and push-on flap protection
  • Unique sample ‘dribble’ feature
Delivery information: Supplied with a soft carrying case and instructions.
Specifications
  • Område:
    45 - 80 °Bx
  • Opløsning:
    0,2 °Bx
  • Beskrivelse:
    Sukker
  • Seq:
    16