Orders LinkContactUsLinkComponent
Trypsin
Trypsin
Trypsin-EDTA 10X, 100 mL
Katalog # X0930-100
Leverandør:   Biowest
Trypsin
Katalog # X0930-100
Leverandør:  Biowest
CAS nummer:  
Trypsin
Trypsin
Trypsin-EDTA 10X, 100 mL
CAS nummer:  
Price
164,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Specifikationer
Specifikationer
  • Pakn:
    100 mL
  • Beskrivelse:
    Trypsin-EDTA 10X