Orders LinkContactUsLinkComponent
Dog serum
Dog serum
Dog serum, Sterile filtered, Frossen
Katalog # S2900-100
Leverandør:   Biowest
Dog serum
Katalog # S2900-100
Leverandør:  Biowest
Dog serum
Dog serum, Sterile filtered, Frossen
Price
Spørg for pris
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
Dog serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Dog serum is used as a blocking agent in immunoassays and controls.
Specifikationer
  • Serum format:
    Frossen
  • Sterilitet:
    Sterile filtered
  • Beskrivelse:
    Dog serum