Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bovine plasma

Bovine plasma

Bovine plasma with sodium citrate, 500 ml
Katalog # S0260-500
Leverandør:   Biowest
Bovine plasma
Katalog # S0260-500
Leverandør:  Biowest
CAS nummer:  
Bovine plasma
Bovine plasma

Bovine plasma with sodium citrate, 500 ml

CAS nummer:  
Price
1.070,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.

Documents

Specifikationer

Specifikationer

  • Pakn:
    500 ml
  • Beskrivelse:
    Bovine plasma with sodium citrate

Specifikationer