Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bovine plasma

Bovine plasma

Bovine plasma with sodium citrate, 100 ml
Katalog # S0260-100
Leverandør:   Biowest
Bovine plasma
Katalog # S0260-100
Leverandør:  Biowest
CAS nummer:  
Bovine plasma
Bovine plasma

Bovine plasma with sodium citrate, 100 ml

CAS nummer:  
Price
258,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.

Documents

Specifikationer

Specifikationer

  • Pakn:
    100 ml
  • Beskrivelse:
    Bovine plasma with sodium citrate

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.