Orders LinkContactUsLinkComponent
Trypsin
Trypsin
Trypsin-EDTA 1X, with sodium chloride, lyophilised, 1 g
Katalog # P0940-1GR
Leverandør:   Biowest
Trypsin
Katalog # P0940-1GR
Leverandør:  Biowest
CAS nummer:  
Trypsin
Trypsin
Trypsin-EDTA 1X, with sodium chloride, lyophilised, 1 g
CAS nummer:  
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Specifikationer
Specifikationer
  • Pakn:
    1 g
  • Beskrivelse:
    Trypsin-EDTA 1X, with sodium chloride, lyophilised