Gentamicin sulfat solution 10 mg/ml
10 mL
Supplier: Biowest
Jump to:
Product Details & Documents
Gentamicin sulfate acts against gram-negative, gram-positive bacteria and mycoplasma.
- Storage temperature of –20 °C
- pH of 4,5±0,3
- Shelf life of 24 months
- Endotoxin: <10 EU/ml (<1 ng/ml)
Used in cell culture applications at 5 ml/l. This concentration is for tissue culture media containing serum, serum-free media generally require lower concentration.
Caution: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.
Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
- CAS Number:1405-41-0
- MDL Number:MFCD00270181
Certificates
Search by Lot NumberSpecifications
Specifications
- Pakn:10 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



